6-Nitro-m-benzoesyre ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Methoxy-2-nitrobenzoic acid

M2245-5G M2245-25G
TCIAM2245-5GEA 222 DKK
TCIAM2245-5G TCIAM2245-25G
6-Nitro-m-benzoesyre ≥97.0% (by GC, titration analysis)
6-Nitro-m-benzoesyre
Formel: C₈H₇NO₅
MDL Number: MFCD00151836
CAS-nummer: 1882-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaddc35c1faa53f1acfbd3840de80f60378f9b58&vid=eaddc35c1faa53f1acfbd3840de80f60378f9b58&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR