2-Methylbenzhydryl chloride ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(alpha-Chlorobenzyl)toluene, Chloro(2-methylphenyl)phenylmethane

M2211-25G M2211-5G
TCIAM2211-25GEA 1260 DKK
TCIAM2211-25G TCIAM2211-5G
2-Methylbenzhydryl chloride ≥98.0% (by GC, titration analysis)
2-Methylbenzhydryl chloride
Formel: C₁₄H₁₃Cl
Kogepunkt: 136°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00972765
CAS-nummer: 41870-52-4
EINECS: 431-450-1
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR