4-Methylcyclohexen

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

M2201-5G M2201-25G
TCIAM2201-5GEA 706 DKK
TCIAM2201-5G TCIAM2201-25G
4-Methylcyclohexen
(±)-4-Methylcyclohexene
Formel: C₇H₁₂
Kogepunkt: 103°C
Flammepunkt: –1°C
MDL Number: MFCD00001573
CAS-nummer: 591-47-9
EINECS: 209-715-4
UN nummer: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eed3d3182ad3c623bb1e40abee99a4c2f5c0101b&vid=eed3d3182ad3c623bb1e40abee99a4c2f5c0101b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC,NMR) min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR