Methyl-3-(4-hydroxyphenyl)propionate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3-(4-Hydroxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxyphenyl)propanoate, Methyl 3-(4-hydroxyphenyl)propionate, 3-(4-Hydroxy-phenyl)-propionic acid-OMe

M2196-25G M2196-5G
TCIAM2196-25GEA 493 DKK
TCIAM2196-25G TCIAM2196-5G
Methyl-3-(4-hydroxyphenyl)propionate ≥98.0% (by GC)
Methyl-3-(4-hydroxyphenyl)propionate
Formel: C₁₀H₁₂O₃
Kogepunkt: 108°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00071577
CAS-nummer: 5597-50-2

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR