4-Methoxy-1-naphthalenemethanol ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
M2086-1G
M2086-5G
TCIAM2086-1GEA
419
DKK
TCIAM2086-1G
TCIAM2086-5G
4-Methoxy-1-naphthalenemethanol ≥96.0% (by GC)
4-Methoxy-1-naphthalenemethanol
Formel:
C₁₂H₁₂O₂ |
MDL Number:
MFCD04089264 CAS-nummer: 16820-54-5 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 75.0 to 80.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...