4-Methoxyphenyl-2-O-acetyl-3-O-allyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M2065-1G
M2065-5G
TCIAM2065-1GEA
936
DKK
TCIAM2065-1G
TCIAM2065-5G
4-Methoxyphenyl-2-O-acetyl-3-O-allyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-2-O-acetyl-3-O-allyl-4,6-O-benzylidene-beta-D-glucopyranoside
Formel:
C₂₅H₂₈O₈ Opbevaringstemperatur: Køleskab |
CAS-nummer:
1477956-18-5 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -21.0 to -25.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...