4-(4-Methoxyphenyl)butyric acid ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M2037-25G M2037-5G
TCIAM2037-25GEA 1010 DKK
TCIAM2037-25G TCIAM2037-5G
4-(4-Methoxyphenyl)butyric acid ≥95.0% (by GC)
4-(4-Methoxyphenyl)butyric acid
Formel: C₁₁H₁₄O₃
Molekylvægt: 194.23 g/mol
Kogepunkt: 196°C
MDL Number: MFCD00004404
CAS-nummer: 4521-28-2
EINECS: 224-849-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 58.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR