4-Methyl-1-phenyl-2-pentanol ≥96.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M2012-25G
M2012-5G
TCIAM2012-25GEA
706
DKK
TCIAM2012-25G
TCIAM2012-5G
4-Methyl-1-phenyl-2-pentanol ≥96.0% (by GC)
4-Methyl-1-phenyl-2-pentanol
Formel:
C₁₂H₁₈O Molekylvægt: 178.28 g/mol Kogepunkt: 123°C |
MDL Number:
MFCD00209515 CAS-nummer: 7779-78-4 EINECS: 231-939-6 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...