Methyl trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester, trans-Methyl 4-hydroxycyclohexanecarboxylate

M1991-5G M1991-1G
TCIAM1991-5GEA 1020 DKK
TCIAM1991-5G TCIAM1991-1G
Methyl trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)
Methyl trans-4-hydroxycyclohexanecarboxylate
Formel: C₈H₁₄O₃
Kogepunkt: 125°C
MDL Number: MFCD20441963
CAS-nummer: 6125-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8043a1f94a626f7857ed45ad6728910a9ff86ad9&vid=8043a1f94a626f7857ed45ad6728910a9ff86ad9&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR