Medroxyprogesterone acetate ≥93.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M1964-1G M1964-5G
TCIAM1964-1GEA 312 DKK
TCIAM1964-1G TCIAM1964-5G
Medroxyprogesterone acetate ≥93.0% (by GC)
Medroxyprogesterone acetate
Formel: C₂₄H₃₄O₄
MDL Number: MFCD00010483
CAS-nummer: 71-58-9
EINECS: 200-757-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Melting point 203.0 to 209.0 deg-C
Specific rotation [a]20/D +45.0 to +51.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR