Mono-2-O-(p-toluenesulphonyl)-γ-cyclodextrin ≥95.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M1957-1G
M1957-200MG
TCIAM1957-1GEA
1380
DKK
TCIAM1957-1G
TCIAM1957-200MG
Mono-2-O-(p-toluenesulphonyl)-γ-cyclodextrin ≥95.0% (by HPLC)
Mono-2-O-(p-toluenesulphonyl)-gamma-cyclodextrin
Formel:
C₅₅H₈₆O₄₂S |
CAS-nummer:
97227-32-2 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Specific rotation [a]20/D | +140 to +150 deg (C=4, DMSO) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...