(S)-1-Methyl-3-pyrrolidinol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (S)-3-Hydroxy-1-methylpyrrolidine, (S)-(+)-1-Methyl-3-pyrrolidinol

M1938-1G M1938-5G
TCIAM1938-1GEA 534 DKK
TCIAM1938-1G TCIAM1938-5G
(S)-1-Methyl-3-pyrrolidinol ≥98.0% (by GC, titration analysis)
(S)-1-Methyl-3-pyrrolidinol
Formel: C₅H₁₁NO
Molekylvægt: 101.15 g/mol
Flammepunkt: 71°C
MDL Number: MFCD03788748
CAS-nummer: 104641-59-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +5.0 to +8.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR