Methyl-3-amino-4-methylthiophene-2-carboxylate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 2-Thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, Methyl 3-amino-4-methylthiophene-2-carboxylate
M1772-25G
M1772-5G
TCIAM1772-25GEA
665
DKK
TCIAM1772-25G
TCIAM1772-5G
Methyl-3-amino-4-methylthiophene-2-carboxylate ≥98.0% (by GC)
Methyl-3-amino-4-methylthiophene-2-carboxylate
Formel:
C₇H₉NO₂S |
MDL Number:
MFCD00051822 CAS-nummer: 85006-31-1 EINECS: 285-060-8 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...