Mifepristone ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: RU 486

M1732-1G M1732-5G
TCIAM1732-1GEA 1090 DKK
TCIAM1732-1G TCIAM1732-5G
Mifepristone ≥98.0% (by HPLC, titration analysis)
Mifepristone
Formel: C₂₉H₃₅NO₂
Opbevaringstemperatur: Køleskab
CAS-nummer: 84371-65-3
Merck Index: 14,06186

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 198.0 deg-C
Specific rotation [a]20/D +138.0 to +144.0 deg (C=0.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR