2-(4-Methoxyphenyl)benzothiazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M1654-5G M1654-1G
TCIAM1654-5GEA 805 DKK
TCIAM1654-5G TCIAM1654-1G
2-(4-Methoxyphenyl)benzothiazole ≥98.0% (by GC)
2-(4-Methoxyphenyl)benzothiazole
Formel: C₁₄H₁₁NOS
MDL Number: MFCD01099069
CAS-nummer: 6265-92-5

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR