4-Methoxyphenyl-2,4,6-tri-O-acetyl-3-O-benzyl-β-D-glucopyranoside ≥96.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M1642-1G
M1642-5G
TCIAM1642-1GEA
1340
DKK
TCIAM1642-1G
TCIAM1642-5G
4-Methoxyphenyl-2,4,6-tri-O-acetyl-3-O-benzyl-β-D-glucopyranoside ≥96.0% (by HPLC)
4-Methoxyphenyl-2,4,6-tri-O-acetyl-3-O-benzyl-beta-D-glucopyranoside
Formel:
C₂₆H₃₀O₁₀ Molekylvægt: 502.52 g/mol Opbevaringstemperatur: Køleskab |
CAS-nummer:
303127-79-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Melting point | 158.0 to 162.0 deg-C |
Specific rotation [a]20/D | -20.0 to -23.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...