Phenylacetaldehyd 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Phenyl acetic aldehyde
A14263.14
A14263.22
A14263.36
A14263.14EA
328
DKK
A14263.14
A14263.22
A14263.36
Phenylacetaldehyd 95%
Phenylacetaldehyd
Purity: 95%
CAS No.: 122-78-1
Molecular Formula: C8H8O
Formula Weight: 120.15
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5350
Boiling Point: 194-196°
Beilstein Reference: 385791
Merck Reference: 14,7265
MDL No.: MFCD00006993
Formel:
C₈H₈O Kogepunkt: 194…196 °C Smeltepunkt: 33…34 °C Massefylde: 1,075 Flammepunkt: 68 °C (154 °F) |
MDL Number:
MFCD00006993 CAS-nummer: 122-78-1 EINECS: 204-574-5 Merck Index: 14,7265 |