Methyltri-n-octylammonium Hydrogen sulphate ≥97.0% (by total nitrogen and titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Tri-n-octylmethylammonium Hydrogen Sulphate, N-Methyl-N,N-dioctyloctan-1-aminium hydrogensulphate
M1457-1G
M1457-5G
TCIAM1457-1GEA
353
DKK
TCIAM1457-1G
TCIAM1457-5G
Methyltri-n-octylammonium Hydrogen sulphate ≥97.0% (by total nitrogen and titration analysis)
Methyltri-n-octylammonium Hydrogen sulphate
Formel:
C₂₅H₅₅NO₄S |
MDL Number:
MFCD03095545 CAS-nummer: 59158-14-4 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...