1-Methyl-1-(m-tolyl)hydrazine ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M1419-25G M1419-5G
TCIAM1419-25GEA 4390 DKK
TCIAM1419-25G TCIAM1419-5G
1-Methyl-1-(m-tolyl)hydrazine ≥97.0% (by titrimetric analysis)
1-Methyl-1-(m-tolyl)hydrazine
Formel: C₈H₁₂N₂
MDL Number: MFCD03844802
CAS-nummer: 72233-91-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white clear liquid
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR