2-Methylcyclopenta[l]phenanthrene ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M1411-1G M1411-100MG
TCIAM1411-1GEA 3530 DKK
TCIAM1411-1G TCIAM1411-100MG
2-Methylcyclopenta[l]phenanthrene ≥97.0% (by GC)
2-Methylcyclopenta[l]phenanthrene
Formel: C₁₈H₁₄
MDL Number: MFCD03844801
CAS-nummer: 121254-39-5

Bestil nu

Specifikationer Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 165.0 to 168.0 deg-C
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR