Sodium-2-methyl-2-propene-1-sulphonate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Methallylsulphonic acid Sodium Salt, Sodium 2-methylprop-2-ene-1-sulphonate, Sodium Methallylsulphonate, 2-Methyl-2-propene-1-sulphonic acid Sodium Salt

M1408-25G M1408-500G
TCIAM1408-25GEA 140 DKK
TCIAM1408-25G TCIAM1408-500G
Sodium-2-methyl-2-propene-1-sulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-2-methyl-2-propene-1-sulphonate
Formel: C₄H₈O₃S
MDL Number: MFCD00065344
CAS-nummer: 1561-92-8
EINECS: 216-341-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR