3,3'-Methylenediindole ≥98.0% (by total nitrogen basis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
DIM, 3,3'-Diindolylmethane
M1399-25G
M1399-5G
TCIAM1399-25GEA
1270
DKK
TCIAM1399-25G
TCIAM1399-5G
3,3'-Methylenediindole ≥98.0% (by total nitrogen basis)
3,3'-Methylenediindole
Formel:
C₁₇H₁₄N₂ Molekylvægt: 246.31 g/mol |
MDL Number:
MFCD00195766 CAS-nummer: 1968-05-4 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 165.0 to 171.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...