3-Methylbenzofuran-2-carboxylic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M1392-1G
M1392-5G
TCIAM1392-1GEA
206
DKK
TCIAM1392-1G
TCIAM1392-5G
3-Methylbenzofuran-2-carboxylic acid ≥98.0% (by titrimetric analysis)
3-Methylbenzofuran-2-carboxylic acid
Formel:
C₁₀H₈O₃ Molekylvægt: 176.17 g/mol |
MDL Number:
MFCD00060511 CAS-nummer: 24673-56-1 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 192.0 to 195.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...