4-(4-Methylphenyl)-4-oxobutyric acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M1377-25G
M1377-5G
TCIAM1377-25GEA
1530
DKK
TCIAM1377-25G
TCIAM1377-5G
4-(4-Methylphenyl)-4-oxobutyric acid ≥98.0% (by GC, titration analysis)
4-(4-Methylphenyl)-4-oxobutyric acid
Formel:
C₁₁H₁₂O₃ |
MDL Number:
MFCD00020541 CAS-nummer: 4619-20-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...