Benzyl-L-(+)-mandelate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-Benzyl 2-hydroxy-2-phenylacetate, L-(+)-Mandelic Acid Benzyl Ester

M1355-25G M1355-5G
TCIAM1355-25GEA 1160 DKK
TCIAM1355-25G TCIAM1355-5G
Benzyl-L-(+)-mandelate ≥98.0% (by GC)
Benzyl-L-(+)-mandelate
Formel: C₁₅H₁₄O₃
Molekylvægt: 242.27 g/mol
MDL Number: MFCD00274325
CAS-nummer: 62173-99-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 105.0 to 109.0 deg-C
Specific rotation [a]20/D +31.0 to +37.0 deg (C=1, CH3CN)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR