Diethyl-D-(+)-malate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: D-(+)-Malic Acid Diethyl Ester, (R)-Diethyl 2-hydroxysuccinate, D-(+)-Apple Acid Diethyl Ester

M1351-25G M1351-5G
TCIAM1351-25GEA 2490 DKK
TCIAM1351-25G TCIAM1351-5G
Diethyl-D-(+)-malate ≥98.0% (by GC)
Diethyl-D-(+)-malate
Formel: C₈H₁₄O₅
Molekylvægt: 190.20 g/mol
MDL Number: MFCD02093483
CAS-nummer: 7554-28-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +10.0 to +13.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR