Diethyl L-(-)-Malate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: L-(-)-Malic Acid Diethyl Ester, L-(-)-Apple Acid Diethyl Ester

M1348-25G M1348-5G
TCIAM1348-25GEA 567 DKK
TCIAM1348-25G TCIAM1348-5G
Diethyl L-(-)-Malate ≥97.0% (by GC)
Diethyl-L-(-)-malate
Formel: C₈H₁₄O₅
Molekylvægt: 190.20 g/mol
MDL Number: MFCD00210119
CAS-nummer: 691-84-9

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Specific rotation [a]20/D -10.0 to -13.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR