Sodium meclofenamate hydrate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Meclofenamic acid Sodium salt, 2-[(2,6-Dichloro-3-methylphenyl)amino]benzoic acid Sodium salt, Sodium 2-((2,6-dichloro-3-methylphenyl)amino)benzoate

M1269-10G M1269-1G
TCIAM1269-10GEA 2520 DKK
TCIAM1269-10G TCIAM1269-1G
Sodium meclofenamate hydrate ≥98.0% (by HPLC, total nitrogen)
Sodium meclofenamate hydrate
Formel: C₁₄H₁₁Cl₂NO₂
Molekylvægt: 318.13 g/mol
MDL Number: MFCD00077376
CAS-nummer: 6385-02-0
EINECS: 228-983-3
Merck Index: 14,05779
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85c225ef51edcefdde46b2794769723241da6918&vid=894b4b612a8348c31205bddc0f9c79c619399c89&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR