2-Mesitylenesulphonyl hydrazide ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,4,6-Trimethylbenzenesulphonohydrazide, 2,4,6-Trimethylbenzenesulphonyl Hydrazide

M1198-25G M1198-5G
TCIAM1198-25GEA 1740 DKK
TCIAM1198-25G TCIAM1198-5G
2-Mesitylenesulphonyl hydrazide ≥97.0% (by titrimetric analysis)
2-Mesitylenesulphonyl hydrazide
Formel: C₉H₁₄N₂O₂S
Molekylvægt: 214.28 g/mol
MDL Number: MFCD00007585
CAS-nummer: 16182-15-3
EINECS: 240-319-4
UN nummer: 1325
ADR: 4.1 III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Potassium bromate Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR