6-Methoxy-4-methylquinoline hydrate ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M1087-1G
M1087-5G
TCIAM1087-1GEA
1510
DKK
TCIAM1087-1G
TCIAM1087-5G
6-Methoxy-4-methylquinoline hydrate ≥98.0% (by GC, titration analysis)
6-Methoxy-4-methylquinoline hydrate
Formel:
C₁₁H₁₁NO Kogepunkt: 158°C |
MDL Number:
MFCD00191547 CAS-nummer: 41037-26-7 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Melting point | 51.0 to 54.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...