4-(Methylthio)toluene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Methylthioanisole, Methyl p-Tolyl sulphide

M0949-10G M0949-25G
TCIAM0949-10GEA 411 DKK
TCIAM0949-10G TCIAM0949-25G
4-(Methylthio)toluene ≥98.0% (by GC)
4-(Methylthio)toluene
Formel: C₈H₁₀S
Molekylvægt: 138.23 g/mol
Kogepunkt: 94°C
Flammepunkt: 85°C
MDL Number: MFCD00008560
CAS-nummer: 623-13-2
EINECS: 210-773-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR