Methylthiomethyl p-Tolyl sulphone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
MT-sulphone, Methylmercaptomethyl p-Tolyl Sulphone, Methyl p-Toluenesulphonylmethyl sulphide
M0875-25G
M0875-5G
TCIAM0875-25GEA
3210
DKK
TCIAM0875-25G
TCIAM0875-5G
Methylthiomethyl p-Tolyl sulphone ≥98.0% (by GC)
Methylthiomethyl p-Tolyl sulphone
Formel:
C₉H₁₂O₂S₂ Molekylvægt: 216.31 g/mol |
MDL Number:
MFCD00010750 CAS-nummer: 59662-65-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 85.0 deg-C |
Solubility in Acetone | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...