Maltitol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2S,3R,4R,5R)-4-(((2R,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexane-1,2,3,5,6-pentaol

M0797-25G M0797-500G M0797-100G
TCIAM0797-25GEA 238 DKK
TCIAM0797-25G TCIAM0797-500G TCIAM0797-100G
Maltitol ≥98.0% (by GC)
Maltitol
Formel: C₁₂H₂₄O₁₁
MDL Number: MFCD00006600
CAS-nummer: 585-88-6
EINECS: 209-567-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d59488ccdb416558e4c55d41e46ea89150afc4e7&vid=d59488ccdb416558e4c55d41e46ea89150afc4e7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C
Specific rotation [a]20/D +105 to +109 deg (C=10, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR