10-Methyl-9(10H)-acridone ≥98.0% (by total nitrogen basis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
N-Methyl-9-acridone, 10-Methylacridin-9(10H)-one
M0792-25G
M0792-5G
TCIAM0792-25GEA
3210
DKK
TCIAM0792-25G
TCIAM0792-5G
10-Methyl-9(10H)-acridone ≥98.0% (by total nitrogen basis)
10-Methyl-9(10H)-acridone
Formel:
C₁₄H₁₁NO Molekylvægt: 209.25 g/mol |
MDL Number:
MFCD00005024 CAS-nummer: 719-54-0 EINECS: 211-948-1 |
Specifikationer Testresultater
Appearance | White to Light yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 202.0 to 207.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...