γ-Methyl-γ-decanolactone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M0786-25ML M0786-500ML
TCIAM0786-25MLEA 288 DKK
TCIAM0786-25ML TCIAM0786-500ML
γ-Methyl-γ-decanolactone ≥98.0% (by GC)
gamma-Methyl-gamma-decanolactone
Formel: C₁₁H₂₀O₂
Kogepunkt: 160°C
MDL Number: MFCD00036642
CAS-nummer: 7011-83-8
EINECS: 230-291-1

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR