Methyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
M0368-25G
M0368-250G
TCIAM0368-25GEA
279
DKK
TCIAM0368-25G
TCIAM0368-250G
Methyl-ɑ-D-mannopyranoside ≥98.0% (by HPLC)
Methyl-alpha-D-mannopyranoside
Formel:
C₇H₁₄O₆ |
MDL Number:
MFCD00063262 CAS-nummer: 617-04-9 EINECS: 210-502-3 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 192.0 to 196.0 deg-C |
Specific rotation [a]20/D | +77.0 to +82.0 deg (C=4, H2O) |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...