5-Methyl-1,10-phenanthrolin for colorimetric determination of iron

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

M0300-1G M0300-100MG
TCIAM0300-1GEA 1500 DKK
TCIAM0300-1G TCIAM0300-100MG
5-Methyl-1,10-phenanthrolin for colorimetric determination of iron
5-Methyl-1,10-phenanthroline
Formel: C₁₃H₁₀N₂
MDL Number: MFCD00004982
CAS-nummer: 3002-78-6
EINECS: 221-099-9

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Melting point 111.0 to 114.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR