Benzyl methacrylate ≥98.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methacrylic Acid Benzyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M0279-25ML M0279-500ML
TCIAM0279-25MLEA 140 DKK
TCIAM0279-25ML TCIAM0279-500ML
Benzyl methacrylate ≥98.0% (by GC) stabilised
Benzyl methacrylate
Formel: C₁₁H₁₂O₂
Molekylvægt: 176.22 g/mol
Kogepunkt: 106°C
Flammepunkt: 115°C
MDL Number: MFCD00053684
CAS-nummer: 2495-37-6
EINECS: 219-674-4

Bestil nu

Specifikationer Testresultater

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR