Metaldehyde ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Metacetaldehyde, Aldehyde (meta), Acetaldehyde Tetramer
M0076-25G
M0076-500G
TCIAM0076-25GEA
148
DKK
TCIAM0076-25G
TCIAM0076-500G
Metaldehyde ≥97.0% (by titrimetric analysis)
Metaldehyde
Formel:
C₈H₁₆O₄ Kogepunkt: 65°C Smeltepunkt: 246°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00134115 CAS-nummer: 108-62-3 EINECS: 203-600-2 UN nummer: 1332 ADR: 4.1,III |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Oxime Formation) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...