Blydiacetat ≥98.0% (by titrimetric analysis) for perovskite precursor

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acetic acid Lead(II) salt

L0315-1G L0315-5G L0315-25G
TCIAL0315-1GEA 132 DKK
TCIAL0315-1G TCIAL0315-5G TCIAL0315-25G
Blydiacetat ≥98.0% (by titrimetric analysis) for perovskite precursor
Blydiacetat
Formel: (H₃CCOO)₂Pb
Molekylvægt: 325.29 g/mol
Smeltepunkt: 280°C
MDL Number: MFCD00012452
CAS-nummer: 301-04-2
EINECS: 206-104-4
UN nummer: 1616
ADR: 6.1 III
Merck Index: 14,05397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c065a95c6489f2edf51190d919d63b7dc336fe9e&vid=c065a95c6489f2edf51190d919d63b7dc336fe9e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR