Bly(II)chlorid for perovskite precursor

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Lead dichloride

L0292-25G L0292-1G L0292-5G
TCIAL0292-25GEA 1480 DKK
TCIAL0292-25G TCIAL0292-1G TCIAL0292-5G
Bly(II)chlorid for perovskite precursor
Bly(II)chlorid
Formel: Cl₂Pb
Molekylvægt: 278.11 g/mol
Kogepunkt: 951°C
Smeltepunkt: 501°C
MDL Number: MFCD00011157
CAS-nummer: 7758-95-4
EINECS: 231-845-5
UN nummer: 2291
ADR: 6.1 III
Merck Index: 14,05404
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55d81cca74da4cfe6b0046b1417750918326207e&vid=55d81cca74da4cfe6b0046b1417750918326207e&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystaline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR