Lercanidipine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Lercanidipine HCl, 3-(1-((3,3-Diphenylpropyl)(methyl)amino)-2-methylpropan-2-yl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride

L0282-200MG L0282-1G
TCIAL0282-200MGEA 534 DKK
TCIAL0282-200MG TCIAL0282-1G
Lercanidipine hydrochloride ≥98.0% (by HPLC, titration analysis)
Lercanidipine hydrochloride
Formel: C₃₆H₄₁N₃O₆·HCl
Molekylvægt: 648.20 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07773089
CAS-nummer: 132866-11-6
Merck Index: 14,05446

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR