Levocetirizine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-[2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride, (R)-2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid dihydrochloride, Levocetirizine 2HCl
L0264-1G
L0264-5G
TCIAL0264-1GEA
247
DKK
TCIAL0264-1G
TCIAL0264-5G
Levocetirizine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Levocetirizine dihydrochloride
Formel:
C₂₁H₂₅ClN₂O₃ |
MDL Number:
MFCD07366507 CAS-nummer: 130018-87-0 Merck Index: 14,02022 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...