Loxoprofen sodium salt dihydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid Sodium Salt, Sodium 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionate

L0252-5G L0252-25G
TCIAL0252-5GEA 509 DKK
TCIAL0252-5G TCIAL0252-25G
Loxoprofen sodium salt dihydrate ≥98.0% (by HPLC, titration analysis)
Loxoprofen sodium salt dihydrate
Formel: C₁₅H₁₈O₃
Smeltepunkt: 198°C
MDL Number: MFCD01745788
CAS-nummer: 226721-96-6
UN nummer: 2811
ADR: 6.1,III
Merck Index: 14,05589
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75cb65553eb780010002a4ac2d11fab913fee980&vid=75cb65553eb780010002a4ac2d11fab913fee980&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 % (calcd.on anh.substance)
Water 11.0 to 13.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR