Lamotrigine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

L0241-1G L0241-5G
TCIAL0241-1GEA 321 DKK
TCIAL0241-1G TCIAL0241-5G
Lamotrigine ≥98.0% (by HPLC, titration analysis)
Lamotrigine
Formel: C₉H₇Cl₂N₅
Molekylvægt: 256.09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00865333
CAS-nummer: 84057-84-1
EINECS: 281-901-8
UN nummer: 2811
ADR: 6.1 III
Merck Index: 14,05353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33cae6c3259a58b93a59e3f4a028144e8eb96bd1&vid=33cae6c3259a58b93a59e3f4a028144e8eb96bd1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR