Levofloxacin Q-acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Levofloxacin carboxylic acid, (S)-9,10-Difluoro-3-methyl-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid, (S)-(-)-9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid

L0238-1G L0238-5G
TCIAL0238-1GEA 608 DKK
TCIAL0238-1G TCIAL0238-5G
Levofloxacin Q-acid ≥98.0% (by HPLC, titration analysis)
Levofloxacin Q-acid
Formel: C₁₃H₉F₂NO₄
Molekylvægt: 281.21 g/mol
MDL Number: MFCD04039905
CAS-nummer: 100986-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a79b4f399b162ce1bc9939f7600c0fbf0b498b8&vid=8a79b4f399b162ce1bc9939f7600c0fbf0b498b8&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -62.0 to -64.0 deg (C=1、DMSO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR