Lanthan(III)oxid

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L0235-25G L0235-500G
TCIAL0235-25GEA 132 DKK
TCIAL0235-25G TCIAL0235-500G
Lanthan(III)oxid
Lanthanum(III) oxide
Formel: La₂O₃
MDL Number: MFCD00011071
CAS-nummer: 1312-81-8
Merck Index: 14,05363
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c74e7931509d6b19df31e8cd763e7c1fcef77f78&vid=c74e7931509d6b19df31e8cd763e7c1fcef77f78&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Subtracting method) min. 99.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR