Lead(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Phthalocyanine Lead(II)

L0230-200MG L0230-1G
TCIAL0230-200MGEA 263 DKK
TCIAL0230-200MG TCIAL0230-1G
Lead(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation
Lead(II) phthalocyanine
Formel: C₃₂H₁₆N₈Pb
MDL Number: MFCD00053951
CAS-nummer: 15187-16-3
UN nummer: 2291
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ebc6a9bedb332728e639e518030e719e06fd976&vid=6c4b9a55a47e26b4e5e02060c91229e021dc7f18&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Dark red to Dark purple to Dark blue powder to crystaline
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR