DL-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: H-DL-Lys-OMe.2HCl, Methyl 2,6-diaminohexanoate dihydrochloride

L0228-25G L0228-5G
TCIAL0228-25GEA 1380 DKK
TCIAL0228-25G TCIAL0228-5G
DL-Lysine methyl ester dihydrochloride ≥98.0% (by total nitrogen basis)
DL-Lysine methyl ester dihydrochloride
Formel: C₇H₁₆N₂O₂·₂HCl
Molekylvægt: 233.13 g/mol
MDL Number: MFCD03547976
CAS-nummer: 34015-48-0
EINECS: 251-794-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -1.0 to +1.0 deg (C=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR