Lomefloxacin hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride

L0216-25G L0216-5G
TCIAL0216-25GEA 1240 DKK
TCIAL0216-25G TCIAL0216-5G
Lomefloxacin hydrochloride ≥98.0% (by HPLC, titration analysis)
Lomefloxacin hydrochloride
Formel: C₁₇H₁₉F₂N₃O₃·HCl
Molekylvægt: 387.82 g/mol
MDL Number: MFCD00214312
CAS-nummer: 98079-52-8
Merck Index: 14,05562

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR